accessibility menu, dialog, popup

UserName

CAS RN 255825-38-8

IUPAC Name: tetrafluoroboranuide 2-{[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}pyridin-1-ium-1-olate
Synonyms: 2-{[(dimethylamino)(dimethyliminio)methyl]sulfanyl}pyridin-1-ium-1-olate tetrafluoroborate
Molecular Weight (g/mol): 313.12
Molecular Formula: C10H16BF4N3OS
InChi Key: LHLFXDQURZVFLK-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)