missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. T35695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35695G 5 g
T356925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35695G Supplier TCI America Supplier No. T35695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 255825-38-8
Molecular Formula C10H16BF4N3OS
Molecular Weight (g/mol) 313.12
MDL Number MFCD05664718
InChI Key LHLFXDQURZVFLK-UHFFFAOYSA-N
Synonym tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate
PubChem CID 10543173
IUPAC Name tetrafluoroboranuide 2-{[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}pyridin-1-ium-1-olate
SMILES F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 5 g
Formula Weight 313.12
Percent Purity ≥98.0% (HPLC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium Tetrafluoroborate
TSCA No
Recommended Storage Freezer (-20 °C)