missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%

Catalog No. p-3737484
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420600050 5 g Glass bottle
AC420600250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420600050 Supplier Thermo Scientific Chemicals Supplier No. 42060 0050
Only null left

CAS: 637-01-4 | C10H16N2·2HCl | 237.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H16N2·2HCl
Molecular Weight (g/mol) 237.17
MDL Number MFCD00012482
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 220°C to 224°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H4[N(CH3)2]2·2HCl
Quantity 5 g
Beilstein 13, 74
Merck Index 10, 9052
Solubility Information Solubility in water: ca 50g/L
Formula Weight 237.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 211-274-8
Recommended Storage Product may darken on storage

RUO – Research Use Only