accessibility menu, dialog, popup

UserName

HHClClH3CNCH3NCH3H3C

CAS RN 637-01-4

IUPAC Name: dihydrogen N1,N1,N4,N4-tetramethylbenzene-1,4-diamine dichloride
Synonyms: dihydrogen wurster's blue dichloride
Molecular Weight (g/mol): 237.17
Molecular Formula: C10H18Cl2N2
InChi Key: FBHKTSXMTASXFJ-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CN(C)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)