missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,4-phenylenediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. T01511G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01511G 1 g
T01515G 5 g
T015125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T01511G Supplier TCI America Supplier No. T01511G

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H18Cl2N2
Molecular Weight (g/mol) 237.168
MDL Number MFCD00012482
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 1 g
Formula Weight 237.17
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,4-phenylenediamine Dihydrochloride