missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,3-propanediamine, 98+%

Catalog No. AAA1950030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1950030 250 g
AAA1950009 10 g
AAA1950018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1950030 Supplier Thermo Scientific Chemicals Supplier No. A1950030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-95-2 | C7H18N2 | 130.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-95-2
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.235
MDL Number MFCD00008337
InChI Key DMQSHEKGGUOYJS-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r
PubChem CID 8084
IUPAC Name N,N,N',N'-tetramethylpropane-1,3-diamine
SMILES CN(C)CCCN(C)C

Specifications

Density 0.82
Boiling Point 145°C to 146°C
Flash Point 31°C (87°F)
Refractive Index 1.4234
Quantity 250 g
UN Number UN2733
Beilstein 1734176
Formula Weight 130.24
Percent Purity 99%
Chemical Name or Material N,N,N',N'-Tetramethyl-1,3-propanediamine
Hazard Category H226-H302-H311+H331-H314-H335
Hazard Statement GHS H Statement
H301-H310-H314-H318-H226-H332
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-818-8
RTECSNumber TX8400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only