accessibility menu, dialog, popup

UserName

CAS RN 110-95-2

IUPAC Name: [3-(dimethylazaniumyl)propyl]dimethylazanium
Synonyms: [3-(dimethylammonio)propyl]dimethylazanium
Molecular Weight (g/mol): 132.25
Molecular Formula: C7H20N2
InChi Key: DMQSHEKGGUOYJS-UHFFFAOYSA-P
SMILES: C[NH+](C)CCC[NH+](C)C
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)