missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,2-diaminopropane 92.0+%, TCI America™

Catalog No. p-7136512
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09475ML 5 mL
T094725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09475ML Supplier TCI America Supplier No. T09475ML

Chemical Identifiers

CAS 1822-45-3
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.235
MDL Number MFCD00014871
InChI Key JUXXCHAGQCBNTI-UHFFFAOYSA-N
PubChem CID 98164
IUPAC Name 1-N,1-N,2-N,2-N-tetramethylpropane-1,2-diamine
SMILES CC(CN(C)C)N(C)C

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 mL
UN Number 2734
Synonym 1,2-Bis(dimethylamino)propane, N,N,N′C,N′C-Tetramethyl-1,2-propanediamine
Formula Weight 130.24
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,2-diaminopropane
EINECSNumber (2)-2378
RTECSNumber TX8350000
TSCA No