accessibility menu, dialog, popup

UserName

CAS RN 1822-45-3

IUPAC Name: [2-(dimethylamino)propyl]dimethylamine
Synonyms: [2-(dimethylamino)propyl]dimethylamine
Molecular Weight (g/mol): 130.24
Molecular Formula: C7H18N2
InChi Key: JUXXCHAGQCBNTI-UHFFFAOYNA-N
SMILES: CC(CN(C)C)N(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results