missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(p-tolyl)benzidine (purified by sublimation) 98.0+%, TCI America™

Catalog No. T32651G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32651G Supplier TCI America Supplier No. T32651G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76185-65-4
Molecular Formula C40H36N2
Molecular Weight (g/mol) 544.742
MDL Number MFCD00799300
InChI Key MVIXNQZIMMIGEL-UHFFFAOYSA-N
Synonym N,N,N′C,N′C-Tetrakis(4-methylphenyl)benzidine
PubChem CID 9850394
IUPAC Name 4-methyl-N-[4-[4-(4-methyl-N-(4-methylphenyl)anilino)phenyl]phenyl]-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=C(C=C5)C)C6=CC=C(C=C6)C

Specifications

Melting Point 218°C
Color White
Quantity 1 g
Formula Weight 544.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetrakis(p-tolyl)benzidine (purified by sublimation)