accessibility menu, dialog, popup

UserName

CAS RN 76185-65-4

CH3NCH3NH3CCH3

CAS RN 76185-65-4

IUPAC Name: N4,N4,N4',N4'-tetrakis(4-methylphenyl)-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4,N4',N4'-tetrakis(4-methylphenyl)-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 544.74
Molecular Formula: C40H36N2
InChi Key: MVIXNQZIMMIGEL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results