missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N″,N″-Pentakis(2-hydroxypropyl)diethylenetriamine 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 g |
| Formula Weight | 393.57 |
| Percent Purity | ≥95.0% (T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | N,N,N′,N′′,N′′-Pentakis(2-hydroxypropyl)diethylenetriamine |
Safety and Handling
| TSCA | Yes |