accessibility menu, dialog, popup

UserName

CAS RN 17121-34-5

IUPAC Name: 1-[bis({2-[bis(2-hydroxypropyl)amino]ethyl})amino]propan-2-ol
Synonyms: 1-[bis({2-[bis(2-hydroxypropyl)amino]ethyl})amino]propan-2-ol
Molecular Weight (g/mol): 393.57
Molecular Formula: C19H43N3O5
InChi Key: LWMWZNOCSCPBCH-UHFFFAOYNA-N
SMILES: CC(O)CN(CCN(CC(C)O)CC(C)O)CCN(CC(C)O)CC(C)O
Grade 
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results