missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. B38171G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38171G 1 g
B38175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38171G Supplier TCI America Supplier No. B38171G
Only null left

Chemical Identifiers

CAS 207683-26-9
Molecular Formula C13H20F6N3O3P
Molecular Weight (g/mol) 411.29
MDL Number MFCD00192457
InChI Key TVWKVGNCIJUXMY-UHFFFAOYSA-N
Synonym hspyu, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate, n,n,n',n'-bis tetramethylene-o-n-succinimidyl uronium hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 2,5-dioxopyrrolidin-1-yl oxy bis pyrrolidin-1-yl methylium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate chn, bis pyrrolidin-1-yl methylidene 2,5-dioxopyrrolidin-1-yl oxidanium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate
PubChem CID 16218611
IUPAC Name 1-{[(2,5-dioxopyrrolidin-1-yl)oxy](pyrrolidin-1-yl)methylidene}-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.O=C1CCC(=O)N1OC(N1CCCC1)=[N+]1CCCC1

Specifications

Melting Point 143°C
Color White
Quantity 1 g
Formula Weight 411.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Bis(tetramethylene)-O-(N-succinimidyl)uronium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator