accessibility menu, dialog, popup

UserName

NNONOOPFFFFFF

CAS RN 207683-26-9

IUPAC Name: 1-{[(2,5-dioxopyrrolidin-1-yl)oxy](pyrrolidin-1-yl)methylidene}-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-{[(2,5-dioxopyrrolidin-1-yl)oxy](pyrrolidin-1-yl)methylidene}-1λ⁵-pyrrolidin-1-ylium hexafluorophosphate
Molecular Weight (g/mol): 411.29
Molecular Formula: C13H20F6N3O3P
InChi Key: TVWKVGNCIJUXMY-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.O=C1CCC(=O)N1OC(N1CCCC1)=[N+]1CCCC1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)