missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Ethylenebisacrylamide 97.0+%, TCI America™
Supplier: TCI America E108625G
Specifications
| N,N′-Ethylenebisacrylamide | |
| 145°C | |
| C8H12N2O2 | |
| 25 g | |
| AYGYHGXUJBFUJU-UHFFFAOYSA-N | |
| N-[2-(prop-2-enoylamino)ethyl]prop-2-enamide | |
| 168964 | |
| ≥97.0% (GC) |
| 2956-58-3 | |
| White-Yellow | |
| MFCD00042935 | |
| n,n'-ethylenebisacrylamide, n,n-ethylenebisacrylamide, ethylene bis acrylamide, n,n'-ethane-1,2-diyl diacrylamide, 2-propenamide, n,n'-1,2-ethanediylbis, n,n'-bis acryloyl-1,2-diaminoethane, n,n'-bis acryloyl-1,2-diaminoethane, stabilized with hydroquinone, n-2-prop-2-enoylamino ethyl prop-2-enamide, n-2-prop-2-enamido ethyl prop-2-enamide, ethylenebisacrylamide | |
| C=CC(=O)NCCNC(=O)C=C | |
| 168.196 | |
| 168.20 | |
| Crystalline Powder |
Chemical Identifiers
| 2956-58-3 | |
| 168.196 | |
| AYGYHGXUJBFUJU-UHFFFAOYSA-N | |
| 168964 | |
| C=CC(=O)NCCNC(=O)C=C |
| C8H12N2O2 | |
| MFCD00042935 | |
| n,n'-ethylenebisacrylamide, n,n-ethylenebisacrylamide, ethylene bis acrylamide, n,n'-ethane-1,2-diyl diacrylamide, 2-propenamide, n,n'-1,2-ethanediylbis, n,n'-bis acryloyl-1,2-diaminoethane, n,n'-bis acryloyl-1,2-diaminoethane, stabilized with hydroquinone, n-2-prop-2-enoylamino ethyl prop-2-enamide, n-2-prop-2-enamido ethyl prop-2-enamide, ethylenebisacrylamide | |
| N-[2-(prop-2-enoylamino)ethyl]prop-2-enamide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator