missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dipropargyl-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. D49645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49645G 5 g
D49641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49645G Supplier TCI America Supplier No. D49645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18773-54-1
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
InChI Key NNPRNGRENISUMK-UHFFFAOYSA-N
PubChem CID 329147
IUPAC Name 4-methyl-N,N-bis(prop-2-ynyl)benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CC#C)CC#C

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 5 g
Formula Weight 247.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dipropargyl-p-toluenesulfonamide