accessibility menu, dialog, popup

UserName

H3CSOONCHCH

CAS RN 18773-54-1

IUPAC Name: 4-methyl-N,N-bis(prop-2-yn-1-yl)benzene-1-sulfonamide
Synonyms: 4-methyl-N,N-bis(prop-2-yn-1-yl)benzenesulfonamide
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: NNPRNGRENISUMK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N(CC#C)CC#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)