missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylurea, 98%

Catalog No. p-7024304
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AAA1872014 25 g
AAA1872022 100 g
2 options

Catalog No. AAA1872014

Supplier: Thermo Scientific Chemicals A1872014

Only null left

CAS: 102-07-8 | C13H12N2O | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-07-8
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003017
InChI Key GWEHVDNNLFDJLR-UHFFFAOYSA-N
Synonym n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea
PubChem CID 7595
ChEBI CHEBI:41320
IUPAC Name 1,3-diphenylurea
SMILES O=C(NC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 240°C to 244°C
Density 1.239
Boiling Point 260°C (decomposition)
Quantity 25 g
Beilstein 782650
Merck Index 14,1785
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material N,N'-Diphenylurea
EINECSNumber 203-003-7
RTECSNumber FD9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only