Learn More
N,N'-Diphenylthiourea, 98%
CAS: 102-08-9 | C13H12N2S | 228.31 g/mol
Supplier: Thermo Scientific Chemicals A1964436
Description
N,N'-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| N,N'-Diphenylthiourea | |
| 150°C to 154°C | |
| C13H12N2S | |
| 500 g | |
| 644277 | |
| thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca | |
| FCSHMCFRCYZTRQ-UHFFFAOYSA-N | |
| 1,3-diphenylthiourea | |
| 700999 | |
| 98% |
| 102-08-9 | |
| 1.32 | |
| MFCD00004921 | |
| UN2811 | |
| 14,3333 | |
| Slightly soluble in water. | |
| S=C(NC1=CC=CC=C1)NC1=CC=CC=C1 | |
| 228.31 | |
| 228.32 |
Chemical Identifiers
| 102-08-9 | |
| 228.31 | |
| FCSHMCFRCYZTRQ-UHFFFAOYSA-N | |
| 700999 | |
| S=C(NC1=CC=CC=C1)NC1=CC=CC=C1 |
| C13H12N2S | |
| MFCD00004921 | |
| thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca | |
| 1,3-diphenylthiourea |
Safety and Handling
GHS H Statement
H300-H317
Fatal if swallowed.
May cause an allergic skin reaction.
P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H361d-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 203-004-2
RTECSNumber : FE1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only