missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylthiourea, 98%

Catalog No. AAA1964422
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1964422 100 g
AAA1964436 500 g
AAA196440E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1964422 Supplier Thermo Scientific Chemicals Supplier No. A1964422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-08-9 | C13H12N2S | 228.31 g/mol

N,N'-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 102-08-9
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00004921
InChI Key FCSHMCFRCYZTRQ-UHFFFAOYSA-N
Synonym thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca
PubChem CID 700999
IUPAC Name 1,3-diphenylthiourea
SMILES S=C(NC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 150°C to 154°C
Density 1.32
Quantity 100 g
UN Number UN2811
Beilstein 644277
Merck Index 14,3333
Solubility Information Slightly soluble in water.
Formula Weight 228.32
Percent Purity 98%
Chemical Name or Material N,N'-Diphenylthiourea
Hazard Category H317-H361d-H373
Hazard Statement GHS H Statement
H300-H317
Fatal if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-004-2
RTECSNumber FE1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only