missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diphenylbenzidine 98.0+%, TCI America™
$128.98 - $390.80
Chemical Identifiers
| CAS | 531-91-9 |
|---|---|
| Molecular Formula | C24H20N2 |
| Molecular Weight (g/mol) | 336.44 |
| MDL Number | MFCD00003016 |
| InChI Key | FDRNXKXKFNHNCA-UHFFFAOYSA-N |
| Synonym | n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine |
| PubChem CID | 68280 |
| IUPAC Name | N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine |
| SMILES | N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1 |
Chemical Identifiers
| 531-91-9 | |
| 336.44 | |
| FDRNXKXKFNHNCA-UHFFFAOYSA-N | |
| 68280 | |
| N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1 |
| C24H20N2 | |
| MFCD00003016 | |
| n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine | |
| N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine |
Specifications
| 531-91-9 | |
| C24H20N2 | |
| 5 g | |
| FDRNXKXKFNHNCA-UHFFFAOYSA-N | |
| N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine | |
| 68280 | |
| ≥98.0% (HPLC,N) | |
| N,N′-Diphenylbenzidine |
| 242°C | |
| MFCD00003016 | |
| n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine | |
| N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1 | |
| 336.44 | |
| 336.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes