missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylbenzidine, 98%

CAS: 531-91-9 | C24H20N2 | 336.44 g/mol

$119.12 - $409.48

Chemical Identifiers

CAS 531-91-9
Molecular Formula C24H20N2
Molecular Weight (g/mol) 336.44
MDL Number MFCD00003016
InChI Key FDRNXKXKFNHNCA-UHFFFAOYSA-N
Synonym n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
PubChem CID 68280
IUPAC Name 4-(4-anilinophenyl)-N-phenylaniline
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1380206
View Documents
Thermo Scientific Chemicals
A1380206
5 g
Each for $119.12
Only null left
 
AAA1380214
View Documents
Thermo Scientific Chemicals
A1380214
25 g
Each for $409.48
Only null left
 
Description

Description

N,N'-Diphenylbenzidine is an indicator suitable for redox titrations, e.g. with dichromate or cerium(IV) sulfate 1,2. It is also used as intermediates in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diphenylbenzidine is an indicator suitable for redox titrations, e.g. with dichromate or cerium(IV) sulfate 1,2. It is also used as intermediates in organic synthesis.

Solubility
Insoluble in water.

Notes
Light Sensitive. Do not store above 23°C. Store away from Strong bases. Strong acids.
Specifications

Chemical Identifiers

531-91-9
336.44
FDRNXKXKFNHNCA-UHFFFAOYSA-N
68280
N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
C24H20N2
MFCD00003016
n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
4-(4-anilinophenyl)-N-phenylaniline

Specifications

531-91-9
Odorless
MFCD00003016
2220993
14,3319
Insoluble in water.
N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
336.44
336.44
N,N'-Diphenylbenzidine
246°C to 250°C
C24H20N2
5 g
Light sensitive
n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
FDRNXKXKFNHNCA-UHFFFAOYSA-N
4-(4-anilinophenyl)-N-phenylaniline
68280
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-521-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only