missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diphenylbenzidine, 97%
CAS: 531-91-9 | C24H20N2 | 336.44 g/mol
$125.40 - $854.13
Chemical Identifiers
| CAS | 531-91-9 |
|---|---|
| Molecular Formula | C24H20N2 |
| Molecular Weight (g/mol) | 336.44 |
| MDL Number | MFCD00003016 |
| InChI Key | FDRNXKXKFNHNCA-UHFFFAOYSA-N |
| Synonym | n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine |
| PubChem CID | 68280 |
| IUPAC Name | 4-(4-anilinophenyl)-N-phenylaniline |
| SMILES | N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC117210050
|
Thermo Scientific Chemicals
117210050 |
5 g | Glass bottle |
N/A
|
|
||||
|
AC117210500
|
Thermo Scientific Chemicals
117210500 |
50 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 531-91-9 | |
| 336.44 | |
| FDRNXKXKFNHNCA-UHFFFAOYSA-N | |
| 68280 | |
| N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1 |
| C24H20N2 | |
| MFCD00003016 | |
| n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine | |
| 4-(4-anilinophenyl)-N-phenylaniline |
Specifications
| 531-91-9 | |
| 100.0 | |
| Brown to Gray | |
| 96% min. (HPLC) | |
| C24H20N2 | |
| 5 g | |
| 15, 3349 | |
| FDRNXKXKFNHNCA-UHFFFAOYSA-N | |
| 4-(4-anilinophenyl)-N-phenylaniline | |
| 68280 | |
| 97% | |
| N, N'-Diphenylbenzidine |
| 96.0 | |
| 246°C to 250°C | |
| Authentic | |
| Glass bottle | |
| MFCD00003016 | |
| 13, 223 | |
| n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine | |
| N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1 | |
| 336.44 | |
| 336.42 | |
| Fine Crystalline Powder |
Safety and Handling
EINECSNumber : 208-521-7
RUO – Research Use Only