missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine 98.0+%, TCI America™

Catalog No. p-7128493
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4834200MG 200 mg
D48341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4834200MG Supplier TCI America Supplier No. D4834200MG
Only null left

Chemical Identifiers

CAS 20441-06-9
Molecular Formula C38H32N2
Molecular Weight (g/mol) 516.688
MDL Number MFCD00799301
InChI Key UNZWWPCQEYRCMU-UHFFFAOYSA-N
Synonym N,N′C-Bis(4-methylphenyl)-N,N′C-diphenylbenzidine, 4,4′C-Bis[N-phenyl-N-(p-tolyl)amino]biphenyl
PubChem CID 640095
IUPAC Name 4-methyl-N-[4-[4-(N-(4-methylphenyl)anilino)phenyl]phenyl]-N-phenylaniline
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=C(C=C6)C

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 200 mg
Formula Weight 516.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-N,N′-di(p-tolyl)benzidine