accessibility menu, dialog, popup

UserName

CH3NNH3C

CAS RN 20441-06-9

IUPAC Name: N4,N4'-bis(4-methylphenyl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(4-methylphenyl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 516.69
Molecular Formula: C38H32N2
InChi Key: UNZWWPCQEYRCMU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results