missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diphenyl-N,N'-di(m-tolyl)benzidine (purified by sublimation) 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 170°C |
| Color | White |
| Quantity | 5 g |
| UN Number | 3077 |
| Synonym | N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine, 4,4′-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, TPD |
| Formula Weight | 516.69 |
| Percent Purity | ≥99.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′-Diphenyl-N,N′-di(m-tolyl)benzidine (purified by sublimation) |
Safety and Handling
| EINECSNumber | (4)-1459 |
| TSCA | Yes |