accessibility menu, dialog, popup

UserName

CH3NNH3C

CAS RN 65181-78-4

IUPAC Name: N4,N4'-bis(3-methylphenyl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(3-methylphenyl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 516.69
Molecular Formula: C38H32N2
InChi Key: OGGKVJMNFFSDEV-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC(C)=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)