missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-N,N'-di(m-tolyl)benzidine (purified by sublimation) 99.0+%, TCI America™

Catalog No. D32361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32361G 1 g
D32365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32361G Supplier TCI America Supplier No. D32361G
Only null left

Chemical Identifiers

CAS 65181-78-4
Molecular Formula C38H32N2
Molecular Weight (g/mol) 516.688
MDL Number MFCD00144965
InChI Key OGGKVJMNFFSDEV-UHFFFAOYSA-N
PubChem CID 103315
IUPAC Name 3-methyl-N-[4-[4-(N-(3-methylphenyl)anilino)phenyl]phenyl]-N-phenylaniline
SMILES CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=CC(=C6)C

Specifications

Melting Point 170°C
Color White
Quantity 1 g
UN Number 3077
Synonym N,N′C-Bis(3-methylphenyl)-N,N′C-diphenylbenzidine, 4,4′C-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, TPD
Formula Weight 516.69
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-N,N′-di(m-tolyl)benzidine (purified by sublimation)
EINECSNumber (4)-1459
TSCA Yes