missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylurea, 98%

Catalog No. AAL0506509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0506509 10 g
AAL0506518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0506509 Supplier Thermo Scientific Chemicals Supplier No. L0506509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 598-94-7 | C3H8N2O | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-94-7
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00007959
InChI Key YBBLOADPFWKNGS-UHFFFAOYSA-N
Synonym n,n-dimethylurea, urea, n,n-dimethyl, asym-dimethylurea, urea, dimethyl, urea, 1,1-dimethyl, n,n-dimethylharnstoff german, n,n-dimethylharnstoff, dimethyl urea, 1.1-dimethylurea, 1,1-dimethyl-urea
PubChem CID 11737
IUPAC Name 1,1-dimethylurea
SMILES CN(C)C(=O)N

Specifications

Melting Point 178°C to 183°C
Density 1.255
Quantity 10 g
Beilstein 1740666
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material N,N-Dimethylurea
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 209-957-0
RTECSNumber YS9867985
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only