accessibility menu, dialog, popup

UserName

CAS RN 598-94-7

IUPAC Name: 1,1-dimethylurea
Synonyms: 1,1-dimethylurea
Molecular Weight (g/mol): 88.11
Molecular Formula: C3H8N2O
InChi Key: YBBLOADPFWKNGS-UHFFFAOYSA-N
SMILES: CN(C)C(N)=O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)