missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylsulfamide, Thermo Scientific Chemicals

Catalog No. AC464680010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464680010 1 g
AC464680050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464680010 Supplier Thermo Scientific Chemicals Supplier No. 464680010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3984-14-3
Molecular Formula C2H8N2O2S
Molecular Weight (g/mol) 124.16
InChI Key QMHAHUAQAJVBIW-UHFFFAOYSA-N
IUPAC Name (dimethylsulfamoyl)amine
SMILES CN(C)S(N)(=O)=O

Specifications

Melting Point 97.0°C
Quantity 1 g
Formula Weight 124.16
Chemical Name or Material N,N-Dimethylsulfamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only