accessibility menu, dialog, popup

UserName

H3CNCH3SNH2OO

CAS RN 3984-14-3

IUPAC Name: (dimethylsulfamoyl)amine
Synonyms: (dimethylsulfamoyl)amine
Molecular Weight (g/mol): 124.16
Molecular Formula: C2H8N2O2S
InChi Key: QMHAHUAQAJVBIW-UHFFFAOYSA-N
SMILES: CN(C)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)