missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylsulfamide 98.0+%, TCI America™
$195.73 - $685.97
Chemical Identifiers
| CAS | 3984-14-3 |
|---|---|
| Molecular Formula | C2H8N2O2S |
| Molecular Weight (g/mol) | 124.158 |
| MDL Number | MFCD01861286 |
| InChI Key | QMHAHUAQAJVBIW-UHFFFAOYSA-N |
| Synonym | N,N-Dimethylsulfamoylamide |
| PubChem CID | 134472 |
| ChEBI | CHEBI:83516 |
| IUPAC Name | [methyl(sulfamoyl)amino]methane |
| SMILES | CN(C)S(=O)(=O)N |
Chemical Identifiers
| 3984-14-3 | |
| 124.158 | |
| QMHAHUAQAJVBIW-UHFFFAOYSA-N | |
| 134472 | |
| [methyl(sulfamoyl)amino]methane |
| C2H8N2O2S | |
| MFCD01861286 | |
| N,N-Dimethylsulfamoylamide | |
| CHEBI:83516 | |
| CN(C)S(=O)(=O)N |
Specifications
| 3984-14-3 | |
| White | |
| MFCD01861286 | |
| N,N-Dimethylsulfamoylamide | |
| CN(C)S(=O)(=O)N | |
| 124.158 | |
| CHEBI:83516 | |
| ≥98.0% (N) | |
| N,N-Dimethylsulfamide |
| 97°C | |
| C2H8N2O2S | |
| 1 g | |
| QMHAHUAQAJVBIW-UHFFFAOYSA-N | |
| [methyl(sulfamoyl)amino]methane | |
| 134472 | |
| 124.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No