missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylsulfamide 98.0+%, TCI America™

Catalog No. D26401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26401G 1 g
D26405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26401G Supplier TCI America Supplier No. D26401G

Chemical Identifiers

CAS 3984-14-3
Molecular Formula C2H8N2O2S
Molecular Weight (g/mol) 124.158
MDL Number MFCD01861286
InChI Key QMHAHUAQAJVBIW-UHFFFAOYSA-N
Synonym N,N-Dimethylsulfamoylamide
PubChem CID 134472
ChEBI CHEBI:83516
IUPAC Name [methyl(sulfamoyl)amino]methane
SMILES CN(C)S(=O)(=O)N

Specifications

Melting Point 97°C
Color White
Quantity 1 g
Formula Weight 124.16
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethylsulfamide