missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylsuccinamic acid, 98%

Catalog No. AAL0879614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0879614 25 g
AAL0879606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0879614 Supplier Thermo Scientific Chemicals Supplier No. L0879614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2564-95-6 | C6H11NO3 | 145.158 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2564-95-6
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00043505
InChI Key WAIGPJMZARQZDX-UHFFFAOYSA-N
Synonym n,n-dimethylsuccinamic acid, 4-dimethylamino-4-oxobutanoic acid, nn-dimethylsuccinamic acid, butanoic acid, 4-dimethylamino-4-oxo, n,n-dimethylsuccinic acid monoamide, 3-dimethylcarbamoyl propanoic acid, succinamic acid, n,n-dimethyl, n-dimethyl-succinamic acid, acmc-20ak89, succinamic acid,n-dimethyl
PubChem CID 75717
IUPAC Name 4-(dimethylamino)-4-oxobutanoic acid
SMILES CN(C)C(=O)CCC(=O)O

Specifications

Melting Point 82°C to 87°C
Quantity 25 g
Beilstein 1762788
Solubility Information Slightly soluble in water.
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material N,N-Dimethylsuccinamic acid
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only