accessibility menu, dialog, popup

UserName

CAS RN 2564-95-6

IUPAC Name: 3-(dimethylcarbamoyl)propanoic acid
Synonyms: 3-(dimethylcarbamoyl)propanoic acid
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: WAIGPJMZARQZDX-UHFFFAOYSA-N
SMILES: CN(C)C(=O)CCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results