missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylpiperazine, 98.5%

Catalog No. p-119814
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC116650050 5 mL Glass bottle
AC116651000 100 mL Glass bottle
2 options

Catalog No. AC116650050

Supplier: Thermo Scientific Chemicals 116650050

Only null left

CAS: 106-58-1 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-58-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00006156
InChI Key RXYPXQSKLGGKOL-UHFFFAOYSA-N
Synonym n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
PubChem CID 7818
IUPAC Name 1,4-dimethylpiperazine
SMILES CN1CCN(CC1)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -1°C
Color Colorless to Yellow
Density 0.8520g/mL
Boiling Point 130°C to 133°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4453 to 1.4473
Quantity 5 mL
Beilstein 23, 7
Specific Gravity 0.852
Solubility Information Solubility in water: miscible
Viscosity 1.5 mPa.s (20°C)
Formula Weight 114.19
Percent Purity 98.50%
Physical Form Liquid
Chemical Name or Material N, N'-Dimethylpiperazine, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-412-0
RTECSNumber TL5945000
TSCA TSCA

RUO – Research Use Only