missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctylamine, 97%

CAS: 7378-99-6 | C10H23N | 157.30 g/mol

Supplier:  Thermo Scientific Chemicals 225295000

 View more versions of this product

Catalog No. AC225295000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Dimethyloctylamine
7378-99-6
100.0
0.7650g/mL
65°C
96% min. (C8-chains) (GC)
C10H23N
CH3(CH2)7N(CH3)2
500 mL
0.765
Solubility in water: slightly soluble
CCCCCCCCN(C)C
157.30
157.3
Liquid
97%
96.0
-57.0°C
195.0°C
Authentic
Glass bottle
1.4233 to 1.4253
MFCD00009558
04,I,386
n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
UQKAOOAFEFCDGT-UHFFFAOYSA-N
N,N-dimethyloctan-1-amine
16224
97%

Chemical Identifiers

7378-99-6
157.30
UQKAOOAFEFCDGT-UHFFFAOYSA-N
16224
CCCCCCCCN(C)C
C10H23N
MFCD00009558
n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
N,N-dimethyloctan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contac

GHS Signal Word: Danger

EINECSNumber : 230-939-3

RTECSNumber : RG8075000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only