accessibility menu, dialog, popup

UserName

H3CNCH3CH3

CAS RN 7378-99-6

IUPAC Name: dimethyl(octyl)amine
Synonyms: octyldimethylamine
Molecular Weight (g/mol): 157.3
Molecular Formula: C10H23N
InChi Key: UQKAOOAFEFCDGT-UHFFFAOYSA-N
SMILES: CCCCCCCCN(C)C
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)