missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylmethanesulfonamide, 98+%

Catalog No. AAA1445406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1445406 5 g
AAA1445414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1445406 Supplier Thermo Scientific Chemicals Supplier No. A1445406

CAS: 918-05-8 | C3H9NO2S | 123.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 918-05-8
Molecular Formula C3H9NO2S
Molecular Weight (g/mol) 123.17
MDL Number MFCD00014740
InChI Key WCFDSGHAIGTEKL-UHFFFAOYSA-N
Synonym n,n-dimethyl methanesulfonamide, methanesulfonamide, n,n-dimethyl, nn-dimethylmethanesulfonamide, n,n-dimethylmethanesulphonamide, n,n-dimethyl-methanesulfonamide, timtec-bb sbb007962, dimethyl methylsulfonyl amine, methanesulfonamide,n-dimethyl, wcfdsghaigtekl-uhfffaoysa
PubChem CID 70191
IUPAC Name N,N-dimethylmethanesulfonamide
SMILES CN(C)S(=O)(=O)C

Specifications

Melting Point 46°C to 50°C
Boiling Point 119°C to 120°C (17 mmHg)
Quantity 5 g
Beilstein 1747409
Formula Weight 123.17
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethylmethanesulfonamide
EINECSNumber 213-039-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only