CAS RN 918-05-8
IUPAC Name:
N,N-dimethylmethanesulfonamide
Synonyms:
N,N-dimethylmethanesulfonamide
Molecular Weight (g/mol):
123.17
Molecular Formula:
C3H9NO2S
InChi Key:
WCFDSGHAIGTEKL-UHFFFAOYSA-N
SMILES:
CN(C)S(C)(=O)=O
1
–
1
of
1
results
1
N,N-Dimethylmethanesulfonamide, 98+%
CAS: 918-05-8 Molecular Formula: C3H9NO2S Molecular Weight (g/mol): 123.17 MDL Number: MFCD00014740 InChI Key: WCFDSGHAIGTEKL-UHFFFAOYSA-N Synonym: n,n-dimethyl methanesulfonamide,methanesulfonamide, n,n-dimethyl,nn-dimethylmethanesulfonamide,n,n-dimethylmethanesulphonamide,n,n-dimethyl-methanesulfonamide,timtec-bb sbb007962,dimethyl methylsulfonyl amine,methanesulfonamide,n-dimethyl,wcfdsghaigtekl-uhfffaoysa PubChem CID: 70191 IUPAC Name: N,N-dimethylmethanesulfonamide SMILES: CN(C)S(=O)(=O)C