missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexylamine, 99%

Catalog No. AAA1736422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1736422 100 g
AAA1736406 5 g
AAA1736414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1736422 Supplier Thermo Scientific Chemicals Supplier No. A1736422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4385-04-0 | C8H19N | 129.247 g/mol

N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

Solubility
Miscible with methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.

Chemical Identifiers

CAS 4385-04-0
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009522
InChI Key QMHNQZGXPNCMCO-UHFFFAOYSA-N
Synonym n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t
PubChem CID 78090
IUPAC Name N,N-dimethylhexan-1-amine
SMILES CCCCCCN(C)C

Specifications

Density 0.744
Boiling Point 146°C to 150°C
Flash Point 33°C (92°F)
Refractive Index 1.4136
Quantity 100 g
UN Number UN2733
Solubility Information Miscible with methanol.
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material N,N-Dimethylhexylamine
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 224-491-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only