missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexylamine, 99%

Catalog No. p-3757519
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314740050 5 g Glass bottle
AC314740250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314740050 Supplier Thermo Scientific Chemicals Supplier No. 314740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4385-04-0 | C8H19N | 129.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4385-04-0
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.25
MDL Number MFCD00009522
InChI Key QMHNQZGXPNCMCO-UHFFFAOYSA-N
Synonym n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t
PubChem CID 78090
IUPAC Name N,N-dimethylhexan-1-amine
SMILES CCCCCCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.7440g/mL
Boiling Point 146°C to 150°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4126 to 1.4146
Linear Formula CH3(CH2)5N(CH3)2
Quantity 5 g
Specific Gravity 0.744
Formula Weight 129.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylhexylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 224-491-8

RUO – Research Use Only