missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylguanidine sulfate, 98%

Catalog No. p-117582
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408350100 10 g Plastic bottle
AC408350500 50 g Plastic bottle
AC408351000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC408350100 Supplier Thermo Scientific Chemicals Supplier No. 408350100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 598-65-2 | 0·5 H2SO4 | 272.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-65-2
Molecular Formula 0·5 H2SO4
Molecular Weight (g/mol) 272.32
MDL Number MFCD00013131
InChI Key QSCHFHVDZCPIKX-UHFFFAOYSA-N
Synonym 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1
PubChem CID 69024
IUPAC Name 1,1-dimethylguanidine;sulfuric acid
SMILES CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 280.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH)2NC(NH)NH2)2·H2SO4
Quantity 10 g
Beilstein 04,II,574
Solubility Information Solubility in water: 490g/L (20°C)
Formula Weight 272.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N, N-Dimethylguanidine sulfate, 93%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-946-0
TSCA TSCA

RUO – Research Use Only