Learn More
N,N-Dimethylglycine hydrochloride, 99%
CAS: 2491-06-7 | C4H10ClNO2 | 139.58 g/mol
Supplier: Thermo Scientific Chemicals A1889222
Description
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).
Solubility
Soluble in water (almost transparency).
Notes
Hygroscopic, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| N,N-Dimethylglycine hydrochloride | |
| 189°C to 194°C | |
| MFCD00012610 | |
| 3910578 | |
| 14,3244 | |
| Soluble in water (almost transparency). | |
| CN(C)CC(=O)O.Cl | |
| 139.58 | |
| 139.58 |
| 2491-06-7 | |
| C4H10ClNO2 | |
| 100 g | |
| Hygroscopic | |
| n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride | |
| FKASAVXZZLJTNX-UHFFFAOYSA-N | |
| 2-(dimethylamino)acetic acid;hydrochloride | |
| 75605 | |
| 99% |
Chemical Identifiers
| 2491-06-7 | |
| MFCD00012610 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 219-648-2
RTECSNumber : MB9995000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only