missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine hydrochloride, 99%

CAS: 2491-06-7 | C4H10ClNO2 | 139.58 g/mol

Supplier:  Thermo Scientific Chemicals A1889214

 View more versions of this product

Catalog No. AAA1889214


Only null left
Explore available promotions

Description

Description

N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

Solubility
Soluble in water (almost transparency).

Notes
Hygroscopic, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

N,N-Dimethylglycine hydrochloride
189°C to 194°C
MFCD00012610
3910578
14,3244
Soluble in water (almost transparency).
C1=CC(=CC=C1N=C=O)Br
198.02
139.58
2491-06-7
C7H4BrNO
25 g
Hygroscopic
4-bromophenyl isocyanate, 4-bromophenylisocyanate, p-bromophenyl isocyanate, benzene, 1-bromo-4-isocyanato, p-bromophenylisocyanate, p-bromocarbanil, p-bromophenyl carbonimide, isocyanic acid 4-bromophenyl ester, unii-mgr77m31zw, mgr77m31zw
CZQIJQFTRGDODI-UHFFFAOYSA-N
1-bromo-4-isocyanatobenzene
75609
99%

Chemical Identifiers

2491-06-7
MFCD00012610
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 219-648-2

RTECSNumber : MB9995000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only