missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine Ethyl Ester 98.0+%, TCI America™

Catalog No. D072625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D072625G 25 g
D0726500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D072625G Supplier TCI America Supplier No. D072625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33229-89-9
Molecular Formula C6H14NO2
Molecular Weight (g/mol) 132.18
MDL Number MFCD00009175
InChI Key BGCNBOFPABQGNG-UHFFFAOYSA-O
Synonym n,n-dimethylglycine ethyl ester, ethyl 2-dimethylamino acetate, glycine, n,n-dimethyl-, ethyl ester, dimethylglycine ethyl ester, ethyl n,n-dimethylglycinate, ethyl dimethylamino acetate, ethyl dimethylglycinate, di-me-gly-oet, ethyl n,n-dimethylaminoacetate, n,n-dimethylglycineethylester
PubChem CID 118412
IUPAC Name (2-ethoxy-2-oxoethyl)dimethylazanium
SMILES CCOC(=O)C[NH+](C)C

Specifications

Color Colorless
Boiling Point 150°C to 151°C
Quantity 25 g
UN Number 3272
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylglycine Ethyl Ester
EINECSNumber (2)-3875
RTECSNumber MB9990000
TSCA Yes