accessibility menu, dialog, popup

UserName

CAS RN 33229-89-9

IUPAC Name: (2-ethoxy-2-oxoethyl)dimethylazanium
Synonyms: (2-ethoxy-2-oxoethyl)dimethylazanium
Molecular Weight (g/mol): 132.18
Molecular Formula: C6H14NO2
InChi Key: BGCNBOFPABQGNG-UHFFFAOYSA-O
SMILES: CCOC(=O)C[NH+](C)C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)