missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine, 98+%

CAS: 1118-68-9 | C4H9NO2 | 103.121 g/mol

Supplier:  Thermo Scientific Chemicals A1967014

 View more versions of this product

Catalog No. AAA1967014


Only null left

Description

Description

N,N-Dimethylglycine is used as an athletic performance enhancer and immunostimulant. It is also used in the treatment of autism, epilepsy and mitochondrial disease. It is also employed as a biomarker of protein degradation in chronic obstructive lung disease (COPD) patients. Further, it is used as a substrate to identify, differentiate and characterize amino acid methyltransferase. It plays an important role for the development of glycine-based ionic liquids and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

179°C to 183°C
1118-68-9
C4H9NO2
MFCD00004283
n,n-dimethylglycine, dimethylglycine, 2-dimethylamino acetic acid, dimethylamino acetic acid, n,n-dimethylaminoacetic acid, glycine, n,n-dimethyl, n-methylsarcosine, n,n-dimethyl glycine, unii-7797m4cppa, dmg
FFDGPVCHZBVARC-UHFFFAOYSA-N
2-(dimethylamino)acetic acid
25 g
CHEBI:17724
103.12
Hygroscopic
N,N-Dimethylglycine
6 to 7
(CH3)2NCH2CO2H
14,3244
Soluble in water.
CN(C)CC(=O)O
103.121
673
Odorless
≥98%

Chemical Identifiers

1118-68-9
103.121
FFDGPVCHZBVARC-UHFFFAOYSA-N
673
2-(dimethylamino)acetic acid
C4H9NO2
MFCD00004283
n,n-dimethylglycine, dimethylglycine, 2-dimethylamino acetic acid, dimethylamino acetic acid, n,n-dimethylaminoacetic acid, glycine, n,n-dimethyl, n-methylsarcosine, n,n-dimethyl glycine, unii-7797m4cppa, dmg
CHEBI:17724
CN(C)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 214-267-8

RTECSNumber : MB9865000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only